missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

$104.71 - $296.72

Specifications

CAS 79-09-4
Vapor Pressure 2.4 mmHg (20°C)
Vapor Density 2.55 (vs. air)
Density 0.993 g/mL (at 25°C)
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111020306
View Documents
Merck Emd Millipore
944251MLF
1 mL
Each for $104.71
Only null left
 
111019020
View Documents
Merck Emd Millipore
944255MLF
5 mL
Each for $296.72
Only null left
 
Description

Description

Propionic acid (PA) is a naturally occurring carboxylic acid, which in its pure state exists as a colorless corrosive liquid with an unpleasant odor. It is miscible in water. Industrially it is produced by hydrocarboxylation of ethylene in presence nickel carbonyl as a catalyst. PA has been found to reduce food intake, lower the fatty acids content in plasma and liver, might improve tissue insulin sensitivity and exerts immunosuppressive actions. Propionic acid is an excellent raw material as it is stable, cheap and safe and can even be used as a food additive.
Specifications

Specifications

79-09-4
2.55 (vs. air)
Neat
MFCD00002756
141°C
Soluble in water
≥99.5% (GC)
2.4 mmHg (20°C)
0.993 g/mL (at 25°C)
CH3CH2COOH
-24°C to +23°C
Acid C3; Propanoic acid; Propanyl acid
74.08
Analytical Standard
Videos
SDS
Documents

Documents