missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde 98.0+%, TCI America™

Supplier:  TCI America P049825ML

 View more versions of this product

Catalog No. P049825ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propionaldehyde
-81°C
46°C
MFCD00007020
1275
NBBJYMSMWIIQGU-UHFFFAOYSA-N
propanal
527
58.08
Liquid
123-38-6
Colorless
C3H6O
25 mL
propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
CCC=O
58.08
CHEBI:17153
≥98.0% (GC)

Chemical Identifiers

123-38-6
58.08
NBBJYMSMWIIQGU-UHFFFAOYSA-N
527
propanal
C3H6O
MFCD00007020
propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
CHEBI:17153
CCC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0486

RTECSNumber : UE0350000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions