missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde 98.0+%, TCI America™

Catalog No. p-7134638
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
P049825ML 25 mL
P0498500ML 500 mL
2 options

Catalog No. P049825ML

Supplier: TCI America P049825ML

Only null left

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00007020
InChI Key NBBJYMSMWIIQGU-UHFFFAOYSA-N
Synonym propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
PubChem CID 527
ChEBI CHEBI:17153
IUPAC Name propanal
SMILES CCC=O

Specifications

Melting Point -81°C
Color Colorless
Boiling Point 46°C
Quantity 25 mL
UN Number 1275
Formula Weight 58.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propionaldehyde
EINECSNumber (2)-0486
RTECSNumber UE0350000
TSCA Yes
Recommended Storage Refrigerator