missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187521
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019850 1 mL
111018382 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019850 Supplier Merck Emd Millipore Supplier No. 644091ML
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 123-38-6
Ignition Point 404°F
Vapor Pressure 4.89 psi (20°C)
Vapor Density 2 (vs. air)
Density 0.805 g/mL (at 25°C)
Flash Point -40°C - closed cup
Physical Form Neat
Linear Formula CH3 CH2 CHO
MDL Number MFCD00007020
Melting Point -81°C
Boiling Point 46°C to 50°C
Synonym Propanal
Type Analytical Standard
Quantity 1 mL
Formula Weight 58.08
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less