missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid, 98%

Catalog No. p-3737266
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131500100 10 mL Glass bottle
AC131500250 25 mL Glass bottle
AC131501000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC131500100 Supplier Thermo Scientific Chemicals Supplier No. 131500100
Only null left

CAS: 471-25-0 | C3H2O2 | 70.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 16.0°C
Color Colorless to Brown
Density 1.1300g/mL
Boiling Point 153.0°C to 157.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4315 to 1.4335
Linear Formula HC≡CCO2H
Quantity 10 mL
Specific Gravity 1.13
Merck Index 15, 7935
Solubility Information Solubility in water: miscible
Formula Weight 70.04
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Propiolic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gentl
EINECSNumber 207-437-8

RUO – Research Use Only