Learn More
Propiolic acid, 98+%
CAS: 471-25-0 | C3H2O2 | 70.05 g/mol
Supplier: Thermo Scientific Chemicals A1324514
Description
Propiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropiolic acid is a reagent used in the preparation of transition metal complexes, haloalkyl propiolates and halopropenoates.
Solubility
Miscible with water, ether, ethanol and chloroform.
Notes
Store in cool place. Incompatible with bases, oxidizing agents and reducing agents.
Specifications
| Propiolic acid | |
| 18°C | |
| 102°C (200 mmHg) | |
| C3H2O2 | |
| MFCD00004360 | |
| UN2920 | |
| 14,7824 | |
| Miscible with water,ether,ethanol and chloroform. | |
| OC(=O)C#C | |
| 70.05 | |
| CHEBI:33199 | |
| ≥98% |
| 471-25-0 | |
| 1.136 | |
| 58°C (136°F) | |
| 1.433 | |
| 25 g | |
| 878176 | |
| propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate | |
| UORVCLMRJXCDCP-UHFFFAOYSA-N | |
| prop-2-ynoic acid | |
| 10110 | |
| 70.05 |
Chemical Identifiers
| 471-25-0 | |
| 70.05 | |
| UORVCLMRJXCDCP-UHFFFAOYSA-N | |
| 10110 | |
| prop-2-ynoic acid |
| C3H2O2 | |
| MFCD00004360 | |
| propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate | |
| CHEBI:33199 | |
| OC(=O)C#C |
Safety and Handling
GHS H Statement
H301-H310-H314-H318-H226
Toxic if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301+H311-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 207-437-8
RTECSNumber : UD9300000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only