missing translation for 'onlineSavingsMsg'
Learn More

Propioin 95.0+%, TCI America™

Catalog No. p-7134881
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09745ML 5 mL
P097425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09745ML Supplier TCI America Supplier No. P09745ML
Only null left

Chemical Identifiers

CAS 4984-85-4
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00043572
InChI Key SKCYVGUCBRYGTE-UHFFFAOYSA-N
Synonym 4-Hydroxy-3-hexanone
PubChem CID 95609
ChEBI CHEBI:18351
IUPAC Name 4-hydroxyhexan-3-one
SMILES CCC(C(=O)CC)O

Specifications

Color Yellow
Boiling Point 167°C
Quantity 5 mL
UN Number 1993
Formula Weight 116.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Propioin