missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Propidium Iodide Solution, Calbiochem™,

Supplier:  MilliporeSigma™ 5370605ML

Catalog No. 5370605ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

25535-16-4
Glass Bottle
5 mL
XJMOSONTPMZWPB-UHFFFAOYSA-M
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
104981
Liquid
pH 7.4
C27H34I2N4
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
668.406
CHEBI:51240

Chemical Identifiers

25535-16-4
668.406
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
CHEBI:51240
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
C27H34I2N4
XJMOSONTPMZWPB-UHFFFAOYSA-M
104981
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions