accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
Sign In
Don't have an account?
Create an Account
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Chemicals
Organic compounds
Organoheterocyclic compounds
Quinolines and derivatives
MilliporeSigma™ Propidium Iodide, Calbiochem™,
Structure Search
Print
MilliporeSigma™ Propidium Iodide, Calbiochem™,
Catalog No.
537059100MG
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
Packaging
537059100MG
100 mg
Glass Bottle
537059250MG
250 mg
Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No.
537059100MG
Supplier
MilliporeSigma™
Supplier No.
537059100MG
Sign In
or
Register
to check your price and availability.
$308.37
/
Each
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
25535-16-4
Molecular Formula
C27H34I2N4
Molecular Weight (g/mol)
668.406
InChI Key
XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym
propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID
104981
ChEBI
CHEBI:51240
IUPAC Name
3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES
CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]
Specifications
Color
Red
Packaging
Glass Bottle
Quantity
100 mg
Physical Form
Solid
Product Suggestions
Documents