Learn More
Thermo Scientific Chemicals Propidium iodide, 95%
CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol
Supplier: Thermo Scientific Chemicals 440300010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Propidium iodide | |
| 220.0°C to 225.0°C | |
| Glass bottle | |
| 1 g | |
| Solubility in water: soluble. | |
| CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] | |
| 668.39 | |
| CHEBI:51240 | |
| 95% |
| 25535-16-4 | |
| 94% min. (Argentometry) | |
| C27H34I2N4 | |
| propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide | |
| XJMOSONTPMZWPB-UHFFFAOYSA-M | |
| 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide | |
| 104981 | |
| 668.39 |
Chemical Identifiers
| 25535-16-4 | |
| 668.39 | |
| propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide | |
| CHEBI:51240 | |
| CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-] |
| C27H34I2N4 | |
| XJMOSONTPMZWPB-UHFFFAOYSA-M | |
| 104981 | |
| 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Warning
EINECSNumber : 247-081-
RUO – Research Use Only