missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Propidium Iodide

Catalog No. 5018262
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018262 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5018262 Supplier Invitrogen™ Supplier No. BMS500PI
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Propodium Iodide

Standard reagent used for assessing cell viability and exclusion of non-viable cells in flow cytometry. PI binds to double-stranded DNA, but is excluded from cells with intact plasma membranes.

  • PI should be analyzed in the PE channel when used as a counterstain for Annexin V FITC
  • 7-AAD can be used in place of PI when using Annexin V PE
  • The advantage of 7-AAD over PI is that there is minimal spectral overlap between these emissions
  • Choose Propidium Iodide as a counterstain with Annexin V apoptosis assays

Specifications

Color Red
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Propidium Iodide
Dye Type Cell-Impermeant
Product Line eBioscience
Quantity 1.5 mL
Emission Wavelength 535/617
What is the concentration of eBioscience Propidium Iodide?

eBioscience Propidium Iodide is supplied at a concentration of 0.02 mg/mL and we recommend using 10 µL in a total staining volume of 200 µL.

For Research Use Only. Not for use in diagnostic procedures.