missing translation for 'onlineSavingsMsg'
Learn More

(Propargyloxy)trimethylsilane, 97%

Catalog No. AAL1293614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1293614 25 g
AAL1293606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1293614 Supplier Thermo Scientific Chemicals Supplier No. L1293614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5582-62-7 | C6H12OSi | 128.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5582-62-7
Molecular Formula C6H12OSi
Molecular Weight (g/mol) 128.246
MDL Number MFCD00042844
InChI Key ZZRPJWCNCLSOLR-UHFFFAOYSA-N
Synonym propargyloxytrimethylsilane, propargyloxy trimethylsilane, o-trimethylsilylpropargyl alcohol, silane, trimethyl 2-propynyloxy, trimethyl prop-2-yn-1-yloxy silane, 3-trimethylsiloxy-1-propyne, trimethyl 2-propynyloxy silane, silane, trimethyl 2-propyn-1-yloxy, trimethyl prop-2-ynyloxy silane, 3-trimethylsiloxypropyne
PubChem CID 79696
IUPAC Name trimethyl(prop-2-ynoxy)silane
SMILES C[Si](C)(C)OCC#C

Specifications

Boiling Point 110°C to 111°C
Quantity 25 g
Linear Formula (CH3)3SiOCH2C≡CH
UN Number UN1993
Beilstein 2036793
Flash Point 8°C (46°F)
Formula Weight 128.25
Percent Purity 97%
Refractive Index 1.408
Sensitivity Moisture sensitive
Density 0.833
Chemical Name or Material (Propargyloxy)trimethylsilane
Show More Show Less
Hazard Category H225-H302-H315-H319-H335
Hazard Statement GHS H Statement
H225-H302
Highly flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 226-981-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only