accessibility menu, dialog, popup

UserName

CAS RN 5582-62-7

IUPAC Name: trimethyl(prop-2-yn-1-yloxy)silane
Synonyms: trimethyl(prop-2-yn-1-yloxy)silane
Molecular Weight (g/mol): 128.25
Molecular Formula: C6H12OSi
InChi Key: ZZRPJWCNCLSOLR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results