missing translation for 'onlineSavingsMsg'
Learn More

Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol

Catalog No. AAL1155609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1155609 10 g
AAL1155618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1155609 Supplier Thermo Scientific Chemicals Supplier No. L1155609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13861-22-8 | C7H8O2 | 124.14 g/mol

Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl methacrylate is a monomer used in polymerization reactions. It is associated with its acetylene and under goes polymerization reaction through the reversible addition fragmentation chain transfer (RAFT) process using cyanoisopropyl dithiobenzoate as a reagent.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 13861-22-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00048123
InChI Key PZAWASVJOPLHCJ-UHFFFAOYSA-N
Synonym propargyl methacrylate, 2-propynyl methacrylate, 2-propenoic acid, 2-methyl-, 2-propynyl ester, 2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester, prop-2-yn-1-yl 2-methylprop-2-enoate, prop-2-ynyl methacrylate, propargylmethacrylate, acmc-1bvgj, prop-2-yn-1-yl methacrylate, propargyl methacrylate, stabilized 10g
PubChem CID 83778
IUPAC Name prop-2-ynyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC#C

Specifications

Density 0.986
Boiling Point 149°C to 151°C
Flash Point 48°C (118°F)
Refractive Index 1.45
Quantity 10 g
UN Number UN3272
Beilstein 906850
Solubility Information Immiscible with water.
Formula Weight 124.14
Percent Purity 98%
Chemical Name or Material Propargyl methacrylate, stabilized with 250ppm 4-methoxyphenol
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 237-599-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only