CAS RN 13861-22-8
IUPAC Name:
prop-2-yn-1-yl 2-methylprop-2-enoate
Synonyms:
prop-2-yn-1-yl 2-methylprop-2-enoate
Molecular Weight (g/mol):
124.14
Molecular Formula:
C7H8O2
InChi Key:
PZAWASVJOPLHCJ-UHFFFAOYSA-N
SMILES:
CC(=C)C(=O)OCC#C
1
–
1
of
1
results
1
Promotions Available
Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol
Propargyl methacrylate, 98%, stab. with 200-300ppm 4-methoxyphenol
CAS: 13861-22-8 Molecular Formula: C7H8O2 Molecular Weight (g/mol): 124.14 MDL Number: MFCD00048123 InChI Key: PZAWASVJOPLHCJ-UHFFFAOYSA-N Synonym: propargyl methacrylate,2-propynyl methacrylate,2-propenoic acid, 2-methyl-, 2-propynyl ester,2-propenoic acid, 2-methyl-, 2-propyn-1-yl ester,prop-2-yn-1-yl 2-methylprop-2-enoate,prop-2-ynyl methacrylate,propargylmethacrylate,acmc-1bvgj,prop-2-yn-1-yl methacrylate,propargyl methacrylate, stabilized 10g PubChem CID: 83778 IUPAC Name: prop-2-ynyl 2-methylprop-2-enoate SMILES: CC(=C)C(=O)OCC#C