missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Ether 98.0+%, TCI America™

Catalog No. p-7135522
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
P23351G 1 g
P23355G 5 g
2 options

Catalog No. P23351G

Supplier: TCI America P23351G

Only null left

Chemical Identifiers

CAS 6921-27-3
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.113
MDL Number MFCD00048108
InChI Key HRDCVMSNCBAMAM-UHFFFAOYSA-N
Synonym propargyl ether, dipropargyl ether, 2-propynyl ether, 1-propyne, 3,3'-oxybis, bispropargyl ether, 3-prop-2-yn-1-yloxy prop-1-yne, propargyl-ether, dipropargyl oxide, di 2-propynyl ether, bis 2-propynyl ether
PubChem CID 23349
IUPAC Name 3-prop-2-ynoxyprop-1-yne
SMILES C#CCOCC#C

Specifications

Color Yellow
Boiling Point 117°C
Quantity 1 g
UN Number 3271
Formula Weight 94.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargyl Ether
RTECSNumber UK5950000
TSCA No
Recommended Storage Refrigerator