accessibility menu, dialog, popup

UserName

HCOCH

CAS RN 6921-27-3

IUPAC Name: 3-(prop-2-yn-1-yloxy)prop-1-yne
Synonyms: 2-propynyl ether
Molecular Weight (g/mol): 94.11
Molecular Formula: C6H6O
InChi Key: HRDCVMSNCBAMAM-UHFFFAOYSA-N
SMILES: C#CCOCC#C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)