missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Chloride 97.0+%, TCI America™

Supplier:  TCI America P08105ML

 View more versions of this product

Catalog No. P08105ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Propargyl Chloride
-78°C
57°C
MFCD00000980
3286
LJZPPWWHKPGCHS-UHFFFAOYSA-N
3-chloroprop-1-yne
12221
≥97.0% (GC)
624-65-7
Yellow
C3H3Cl
5 mL
propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
ClCC#C
74.51
74.51
Liquid

Chemical Identifiers

624-65-7
74.51
LJZPPWWHKPGCHS-UHFFFAOYSA-N
12221
ClCC#C
C3H3Cl
MFCD00000980
propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
3-chloroprop-1-yne
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3300

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions