missing translation for 'onlineSavingsMsg'
Learn More

Propargyl Chloride 97.0+%, TCI America™

Catalog No. p-7134781
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P08105ML 5 mL
P081025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P08105ML Supplier TCI America Supplier No. P08105ML

Chemical Identifiers

CAS 624-65-7
Molecular Formula C3H3Cl
Molecular Weight (g/mol) 74.51
MDL Number MFCD00000980
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
PubChem CID 12221
IUPAC Name 3-chloroprop-1-yne
SMILES ClCC#C

Specifications

Melting Point -78°C
Color Yellow
Boiling Point 57°C
Quantity 5 mL
UN Number 3286
Formula Weight 74.51
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Propargyl Chloride
EINECSNumber (2)-3300
TSCA Yes
Recommended Storage Refrigerator