missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80 wt.% solution in toluene, stabilized

Catalog No. p-4509879
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
AcroSeal™ Glass bottle
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131480500 50 mL Glass bottle
AC131482500 250 mL Glass bottle
AC131480010 1 L AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC131480500 Supplier Thermo Scientific Chemicals Supplier No. 131480500
Only null left

CAS: 106-96-7 | C3H3Br | 118.961 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Specifications

Chemical Name or Material Propargyl bromide
Color White
CAS 106-96-7, 108-88-3
Boiling Point 88.0°C to 90.0°C
Molecular Formula C3H3Br
Linear Formula HC≡CCH2Br
Fieser 06,493; 10,332; 06,495
Density 1.3800g/mL
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
Solubility Information Solubility in water: immiscible.
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
SMILES C#CCBr
IUPAC Name 3-bromoprop-1-yne
Molecular Weight (g/mol) 118.961
PubChem CID 7842
Formula Weight 118.96
Percent Purity 73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.4960
Specific Gravity 1.38
Quantity 50 mL
Flash Point 4°C
Physical Form Powder
Show More Show Less

RUO – Research Use Only