Learn More
Propargyl alcohol, 99%
CAS: 107-19-7 | C3H4O | 56.06 g/mol
Supplier: Thermo Scientific Chemicals A102950F
Description
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Specifications
| Propargyl alcohol | |
| -53°C | |
| 114°C to 115°C | |
| C3H4O | |
| HC≡CCH2OH | |
| 2500 mL | |
| 506003 | |
| propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne | |
| TVDSBUOJIPERQY-UHFFFAOYSA-N | |
| prop-2-yn-1-ol | |
| 7859 | |
| 56.06 |
| 107-19-7 | |
| 0.949 | |
| 33°C (91°F) | |
| 1.432 | |
| MFCD00002912 | |
| UN2929 | |
| 14,7809 | |
| Fully miscible in water. | |
| OCC#C | |
| 56.06 | |
| CHEBI:28905 | |
| 99% |
Chemical Identifiers
| 107-19-7 | |
| 56.06 | |
| TVDSBUOJIPERQY-UHFFFAOYSA-N | |
| 7859 | |
| prop-2-yn-1-ol |
| C3H4O | |
| MFCD00002912 | |
| propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne | |
| CHEBI:28905 | |
| OCC#C |
Safety and Handling
GHS H Statement
H301-H311-H331-H314-H226
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H301-H310+H330-H314-H335-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber : 203-471-2
RTECSNumber : UK5075000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only