accessibility menu, dialog, popup

UserName

CAS RN 107-19-7

IUPAC Name: prop-2-yn-1-ol
Synonyms: propargyl alcohol rcra waste number P102 lithium salt propargyl alcohol prop-2-in-1-ol sodium salt propargyl alcohol agrisynth PA propiolic alcohol 3-propynol propargyl alcohol (substance)
Molecular Weight (g/mol): 56.06
Molecular Formula: C3H4O
InChi Key: TVDSBUOJIPERQY-UHFFFAOYSA-N
SMILES: OCC#C
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)