missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propargyl alcohol, 99%
Description
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Specifications
Specifications
| Melting Point | -53°C |
| Density | 0.949 |
| Boiling Point | 114°C to 115°C |
| Flash Point | 33°C (91°F) |
| Refractive Index | 1.432 |
| Linear Formula | HC≡CCH2OH |
| Quantity | 100 mL |
| UN Number | UN2929 |
| Beilstein | 506003 |
| Merck Index | 14,7809 |
| Show More |
Sécurité et manipulation
| Hazard Category | H226-H301-H310+H330-H314-H335-H373 |
| Hazard Statement | GHS H Statement H301-H311-H331-H314-H226 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S. |
| EINECSNumber | 203-471-2 |
| RTECSNumber | UK5075000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only