missing translation for 'onlineSavingsMsg'
Learn More

Propantheline Bromide Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014361
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014361 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014361 Supplier Merck Emd Millipore Supplier No. PHR1575500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Propantheline Bromide belongs to the class of quaternary ammonium compounds and is a synthetic anticholinergic drug. It is widely used as an oral agent and it exhibits antimuscarinic effect in the lower urinary tract.

Specifications

CAS 50-34-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C23H30 NO3 Br
MDL Number MFCD00050291
Synonym Propantheline bromide; (2-Hydroxyethyl)diisopropylmethylammonium bromide xanthene-9-carboxylate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 448.39
Grade Certified Reference Material