missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PRONASE™ Protease, Streptomyces griseus, Nuclease-Free

Catalog No. p-7060185
Change view
Click to view available options
Quantity:
100kU
10kU
25kU
50kU
5kU
Catalog No. Quantity
5370885KU 5kU
53708810KU 10kU
53708825KU 25kU
53708850KU 50kU
537088100KU 100kU
5 options

Catalog No. 5370885KU

Supplier: MilliporeSigma™ 5370885KU

Only null left

Widely used broad-spectrum protease mixture

  • Non-specific protease liquefies mucins and digests proteins to free amino acids
  • Can be used to isolate living chondrocytes
  • Most active at neutral pH, but stable over wide range of pH and temperature

Specifications

Quantity 5kU
Format Lyophilized
pH Optimum 7.5
Content And Storage Store at+2°C to+8°C