missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PRONASE™ Protease, Streptomyces griseus

Catalog No. 5370210KU
Change view
Click to view available options
Quantity:
10kU
250kU
500kU
50kU
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5370210KU 10kU
5370250KU 50kU
53702250KU 250kU
53702500KU 500kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 5370210KU Supplier MilliporeSigma™ Supplier No. 5370210KU

Most active at neutral pH, but stable over wide range of pH and temperature

  • A non-specific protease liquefies mucins and digests proteins to free amino acids
  • Can be used to isolate living chondrocytes

Specifications

Formulation Lyophilized from a solution containing calcium acetate as a stabilizer
Quantity 10kU
Format Lyophilized
pH Optimum 7.5
Content And Storage Store at+2°C to+8°C