missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America P2029100G

 View more versions of this product

Catalog No. P2029100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Promethazine Hydrochloride
226°C
C17H21ClN2S
100 g
promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl
320.879
CHEBI:8462
≥98.0% (HPLC,T)
58-33-3
White-Yellow
MFCD00012652
3077
XXPDBLUZJRXNNZ-UHFFFAOYSA-N
N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
6014
320.88
Crystalline Powder

Chemical Identifiers

58-33-3
320.879
XXPDBLUZJRXNNZ-UHFFFAOYSA-N
6014
N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
C17H21ClN2S
MFCD00012652
promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen
CHEBI:8462
CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SO8225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions