Learn More
Promega x-tracta™ Gel Extractors DFS Item
The x-tracta Gel Extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a 0.13 x 0.33 inch gel piece from agarose gels for easy transfer into a microcentrifuge tube for processing.
Supplier:  Promega A2122
 View more versions of this product
Catalog No. PRA2122
Only null left
Explore available promotions
Description
Description
  • Safe, Convenient Extraction of Gel Slices
  • Removes a 0.13 x 0.33 inch gel piece
  • Disposable device reduces cross-contamination
  • No need to use razor blades or scalpels
Specifications
Specifications
x-tracta™ Gel Extractor
100/Pk.
15°C to 30°C
Product Suggestions
Product Suggestions