Learn More
Promega Wizard SV Gel and PCR Clean-Up System and x-tracta™ Gel Extractor Package
The Wizard™ SV Gel and PCR Clean-Up System is designed to purify DNA fragments of 100bp-10kb from standard or low-melting agarose gels or to purify products directly from PCR or restriction digests.
Supplier: Promega A9283
Description
- Purifies DNA Fragments from Gels or PCR
- Purifies 100bp to 10kb DNA fragments
- Includes gel extractors for added speed and convenience
Specifications
| Wizard™ SV Gel and PCR Clean up System and x-tracta™ Gel Extractor Package | |
| 50 Preparation/25 Extractors | |
| 22°C to 25°C |