missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Unmethylated Lambda DNA
Specifications
Specifications
| Product Type | Lambda DNA |
| Source | Infected GM119 |
| Content And Storage | -30°C to -10°C |
| Quantity | 250 μg |
| For Use With (Application) | Restriction Enzymes sensitive to DNA Methylation |