Learn More
Promega Ultra Pure Deoxynucleotide Triphosphates (dNTPs)
High-quality dNTPs. Provided at a concentration of 100mM in water at pH 7.5.
Supplier: Promega U1211
Description
- High-Quality Nucleotides for Your Workflow
- Greater than 99% triphosphate content
- 100mM in water at pH 7.5
Specifications
| 7.5 | |
| -30°C to -10°C | |
| >99% | |
| cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols |
| dGTP | |
| 100 mM | |
| 40 μm |