Learn More
Promega TNT™ Quick Coupled Transcription/Translation System DFS Item
The TNT Quick Systems are convenient single-tube, coupled transcription/translation reactions for eukaryotic cell-free protein expression.
Supplier:  Promega L2081
 View more versions of this product
Catalog No. PR-L2081
Only null left
Explore available promotions
Description
Description
  • Single-Tube, Coupled Transcription/Translation for Eukaryotic Cell-Free Protein Expression
  • Functional protein is expressed in one hour, ready for use without further purification
  • Use protein for protein:protein and protein:nucleic acid interactions, protein modification and more
  • Choice of SP6 or T7 promoter
  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents.
  • U.S. Patents 5,324,637 and 5,492,817; Australian Patent 660329 and other patents.
  • U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
  • For Laboratory Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patents 6,027,913 and 6,361,949.
Specifications
Specifications
<-65°C
200μL TNT SP6 Quick Master Mix, 5μg Luciferase SP6 Control DNA, 50μL Methionine
TNT SP6
GST pull-downs, Gel shift assays, Co-immunoprecipitation, Characterization of protein modifications, Protein-RNA interactions, Protein activity studies, Confirmation of gene products, Protein truncation test (PTT)
5
Product Suggestions
Product Suggestions