missing translation for 'onlineSavingsMsg'
Learn More

Promega TNT™ Quick Coupled Transcription/Translation System DFS Item

The TNT Quick Systems are convenient single-tube, coupled transcription/translation reactions for eukaryotic cell-free protein expression.

$372.88 - $1,367.67

Specifications

Content And Storage <-65°C
For Use With (Application) GST pull-downs, Gel shift assays, Co-immunoprecipitation, Characterization of protein modifications, Protein-RNA interactions, Protein activity studies, Confirmation of gene products, Protein truncation test (PTT)
View More Specs

Products 4
Catalog Number Mfr. No. Product Type No. of Reactions Price Quantity  
Catalog Number Mfr. No. Product Type No. of Reactions Price Quantity  
PR-L1170 DFS Item
View Documents
Promega
L1170
TNT T7 40 N/A
Only null left
 
PR-L1171 DFS Item
View Documents
Promega
L1171
TNT T7 5 N/A
Only null left
 
PR-L2080 DFS Item
View Documents
Promega
L2080
TNT SP6 40 N/A
Only null left
 
PR-L2081 DFS Item
View Documents
Promega
L2081
TNT SP6 5 N/A
Only null left
 
Description

Description

  • Single-Tube, Coupled Transcription/Translation for Eukaryotic Cell-Free Protein Expression
  • Functional protein is expressed in one hour, ready for use without further purification
  • Use protein for protein:protein and protein:nucleic acid interactions, protein modification and more
  • Choice of SP6 or T7 promoter

  • U.S. Patents 5,641,641; 5,650,289; and 5,814,471; Australian Patent 649289 and other patents.
  • U.S. Patents 5,324,637 and 5,492,817; Australian Patent 660329 and other patents.
  • U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
  • For Laboratory Use. Any use of the product for diagnostics requiring clearance or approval by the FDA may require a license under Mayo Clinic U.S. Patents 6,027,913 and 6,361,949.

Specifications

Specifications

<-65°C
GST pull-downs, Gel shift assays, Co-immunoprecipitation, Characterization of protein modifications, Protein-RNA interactions, Protein activity studies, Confirmation of gene products, Protein truncation test (PTT)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'