missing translation for 'onlineSavingsMsg'
Learn More

Promega TMB One Solution

Catalog No. PRG7431
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG7431 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG7431 Supplier Promega Supplier No. G7431

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TMB One Solution is a chromagen substrate, 3,3',5,5'-tetramethylbenzidine (TMB) provided in a mildly acidic, nonhazardous buffer for HRP detection in ELISA format.

  • Substrate for Horseradish Peroxidase Detection in ELISAs
  • Single solution provided ready-to-use; just add, incubate, stop and read
  • Low background provides greater assay sensitivity

Specifications

For Use With (Application) ELISA
Quantity 100 mL
pH 3.5
Stability 12 Months
Content And Storage 2°C to 10°C
Product Type TMB One Solution
Form Liquid