missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega RNA Lysis Buffer
Description
- Reagent Used With RNA Purification Systems
- RNA Lysis Buffer (RLA) is used to lyse cells during RNA purification
- Component of Promega RNA purification kits available for purchase separately
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Application) | Lysis of cells or tissue |
| Quantity | 50 mL |