Learn More
Promega ReliaPrep™ FFPE Total RNA Miniprep System DFS Item
The ReliaPrep FFPE Total RNA Miniprep System provides a complete, all-inclusive method for purification of quality total RNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.
Supplier:  Promega Z1002
 View more versions of this product
Catalog No. PRZ1002
Only null left
Description
Description
  • Intact, Amplifiable Total RNA from FFPE Tissues
  • Minimal preparation time
  • Safely deparaffinize without harsh organic solvents
  • No overnight digestion
Specifications
Specifications
15 to 30°C
50mL mineral oil, 20mL lysis buffer, 1.1mL proteinase K solution, 2 x 750μL MnCl2, 3.25mL BL buffer, 30mL wash solution, 4 Vial DNase I (lyophilized), 700μL RNase buffer, 13mL nuclease-free water
100 Reactions
Purification of total rna from formalin-fixed paraffin-embedded tissue
Ffpe Total Rna Miniprep System
Product Suggestions
Product Suggestions