missing translation for 'onlineSavingsMsg'
Learn More

Promega ReliaPrep™ FFPE Total RNA Miniprep System

Catalog No. p-4265711
Encompass
Change view
Click to view available options
Quantity:
10 Reactions
100 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRZ1001 10 Reactions
PRZ1002 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRZ1001 Supplier Promega Supplier No. Z1001
Only null left

The ReliaPrep FFPE Total RNA Miniprep System provides a complete, all-inclusive method for purification of quality total RNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.

  • Intact, Amplifiable Total RNA from FFPE Tissues
  • Minimal preparation time
  • Safely deparaffinize without harsh organic solvents
  • No overnight digestion

Specifications

Content And Storage 15 to 30°C
For Use With (Application) Purification of total rna from formalin-fixed paraffin-embedded tissue
Includes 5mL mineral oil, 1mL lysis buffer, 250μL proteinase K solution, 250μL MnCl2, 3.25mL BL buffer, 3mL wash solution, 1 Vial DNase I (lyophilized), 70μL RNase buffer, 1.25mL nuclease-free
Product Type Ffpe Total Rna Miniprep System
Quantity 10 Reactions