missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega ReliaPrep™ FFPE gDNA Miniprep System
The ReliaPrep FFPE gDNA Miniprep System provides a complete, all-inclusive method for purifying quality genomic DNA from formalin-fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion.
$263.52 - $1,129.15
Specifications
| Content And Storage | 15 to 30°C |
|---|---|
| For Use With (Application) | Purifying genomic dna from formalin-fixed paraffin-embedded tissue |
| Product Type | Ffpe Gdna Miniprep System |
Description
- Intact, Amplifiable DNA from FFPE Tissue
- Minimal preparation time: simply add ethanol and go!
- Safely deparaffinize without using harsh organic solvents
- No overnight digestion required
Specifications
| 15 to 30°C | |
| Purifying genomic dna from formalin-fixed paraffin-embedded tissue | |
| Ffpe Gdna Miniprep System |