missing translation for 'onlineSavingsMsg'
Learn More

Promega Random and Oligo (dT) Primers

Catalog No. p-2776090
Encompass
Change view
Click to view available options
For Use With (Application):
cDNA Synthesis and Cloning
cDNA Synthesis with a Reverse Transcriptase
Primer:
Oligo (dT)15
Random
Product Type:
Oligo (dT)15 Primer
Random Primer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Primer Product Type
PRC1101 cDNA Synthesis with a Reverse Transcriptase Oligo (dT)15 Oligo (dT)15 Primer
PRC1181 cDNA Synthesis and Cloning Random Random Primer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRC1101 Supplier Promega Supplier No. C1101
Only null left

Use for first-strand cDNA synthesis with a reverse transcriptase.

  • Streamline Universal cDNA Synthesis
  • High-quality DNA hexamers of randomized sequence

Specifications

pH 7
Product Type Oligo (dT)15 Primer
Content And Storage -30°C to -10°C
Primer Oligo (dT)15
Quantity 20 μg
For Use With (Application) cDNA Synthesis with a Reverse Transcriptase
Form Liquid