missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega QuantiLum™ Recombinant Luciferase
Description
- Reliable and Dependable Recombinant Luciferase
- Available in bulk for large orders
- Custom options available
- Supplied at a concentration of 10-15mg/ml
The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.
Specifications
Specifications
| Quantity | 1 mg |
| Content And Storage | <-65°C |
| Concentration | 10 to 15 mg/mL |