missing translation for 'onlineSavingsMsg'
Learn More

Promega QuantiLum™ Recombinant Luciferase

Catalog No. PRE1701
Encompass
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE1701 1 mg
PRE1702 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRE1701 Supplier Promega Supplier No. E1701
Only null left

QuantiLum Recombinant Luciferase is a luciferase expressed from a cloned gene from the North American firefly (Photinus pyralis).

  • Reliable and Dependable Recombinant Luciferase
  • Available in bulk for large orders
  • Custom options available
  • Supplied at a concentration of 10-15mg/ml

The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Quantity 1 mg
Content And Storage <-65°C
Concentration 10 to 15 mg/mL