Learn More
Promega pSP72 Vector
Used as standard cloning vector or for transcription of RNA in vitro.
Supplier: Promega P2191
Description
- Standard Cloning Vector Used to Transcribe RNA In Vitro
- Versatile vector that can be used for standard cloning and in vitro transcription
- SP6 and T7 RNA polymerase promoters flank the multiple cloning region
- Multiple cloning site provides a convenient selection of restriction sites for cloning
Spécifications
| SP6, T7 | |
| -30°C to -10°C | |
| pSP72 vector |
| XhoI, PvuII, HindIII, SphI, PstI, SalI, AccI, XbaI, BamHI, SmaI, KpnI, SacI, EcoRI, ClaI, EcoRV and BglII | |
| 20 μg | |
| Cloning and transcription of RNA in-vitro |