Learn More
Promega psiCHECK™ Vectors DFS Item
psiCHECK-1 and psiCHECK-2 Vectors enable monitoring of changes in expression of a target gene fused to a reporter gene. They are used for optimizing RNA interference assays.
Supplier:  Promega C8021
Catalog No. PR-C8021
Only null left
Explore available promotions
Description
Description
  • Vectors for Optimizing RNA Interference Assays
  • Monitor changes in expression of a target gene fused to a reporter gene; decreases in luciferase activity correlate with RNAi
  • psiCHECK™-1 Vector is used to monitor RNAi effects in live cells
  • psiCHECK™-2 Vector designed for endpoint lytic assays
Specifications
Specifications
Renilla Luciferase, Firefly Luciferase
20 μg
Provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi), monitors changes in expression of target gene
-30°C to -10°C
psiCHECK-2 vector
Product Suggestions
Product Suggestions