missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega pRL Family of Renilla Luciferase Control Reporter Vectors
Description
- Four Vectors that Contain Wild-Type Renilla Luciferase for Normalization in Reporter Assays
- A T7 promoter is located immediately upstream of Rluc for in vitro synthesis of Renilla luciferase
- The SV40 late poly(A) signal sequence is positioned downstream of Rluc to provide efficient transcription termination and mRNA polyadenylation
- A prokaryotic origin of replication and β-lactamase gene allow selected propagation of the pRL vectors in E. coli host strains
Specifications
Specifications
| Reporter Gene | Luciferase |
| Storage Buffer | TE Buffer (pH 7.4) |
| Content And Storage | -30°C to -10°C |
| Quantity | 20 μg |
| Vector | pRL-null vector |
| For Use With (Application) | Provide constitutive expression of Renilla reniformis (sea pansy) Luciferase, can be used in combination with a firefly Luciferase vector to co-transfect mammalian cells |
Licensed from University of Georgia Research Foundation, Inc., under U.S. Patent 5,418,155; Canadian Patent 2,105,984 and related patents.