Learn More
Promega pRL Family of Renilla Luciferase Control Reporter Vectors
Catalog No. PRE2241
Encompass
Change view
Click to view available options
Vector:
pRL-CMV vector
pRL-null vector
pRL-SV40 vector
pRL-TK vector
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PRE2241 pRL-TK vector
PRE2271 pRL-null vector
PRE2261 pRL-CMV vector
PRE2231 pRL-SV40 vector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRE2241 Supplier Promega Supplier No. E2241
Only null left
The pRL Vectors are wildtype Renilla luciferase control reporter vectors, which provide constitutive expression of Renilla luciferase and can be used in combination with a firefly luciferase vector to cotransfect mammalian cells.
  • Four Vectors that Contain Wild-Type Renilla Luciferase for Normalization in Reporter Assays
  • A T7 promoter is located immediately upstream of Rluc for in vitro synthesis of Renilla luciferase
  • The SV40 late poly(A) signal sequence is positioned downstream of Rluc to provide efficient transcription termination and mRNA polyadenylation
  • A prokaryotic origin of replication and β-lactamase gene allow selected propagation of the pRL vectors in E. coli host strains
Specifications
Promoter HSV-TK
Reporter Gene Luciferase
Storage Buffer TE Buffer (pH 7.4)
Content And Storage -30°C to -10°C
Quantity 20 μg
Vector pRL-TK vector
For Use With (Application) Provide constitutive expression of Renilla reniformis (sea pansy) Luciferase, can be used in combination with a firefly Luciferase vector to co-transfect mammalian cells
Licensed from University of Georgia Research Foundation, Inc., under U.S. Patent 5,418,155; Canadian Patent 2,105,984 and related patents.