missing translation for 'onlineSavingsMsg'
Learn More

Promega PolyATtract™ System 1000 Magnetic Separation Stand

Catalog No. PRZ5410
Encompass
Change view
Click to view available options
Quantity:
1 Stand
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRZ5410 1 Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRZ5410 Supplier Promega Supplier No. Z5410

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The PolyATtract(R) System 1000 Magnetic Separation Stand is designed for use with the PolyATtract(R) System 1000 and is ideal for applications requiring multiple paramagnetic isolations of biomolecules.

  • Use with the PolyATtract(R) System 1000
  • Contains a strong rare earth magnet
  • Can be used with both 15mL and 50mL tubes

U.S. Patent 5,693,784.

Specifications

Content And Storage 15°C to 30°C
For Use With (Application) In-vitro translation, cDNA synthesis, PCR analysis, RNase (RNase) protection assays, primer extension and northern blots, molecular biology applications, amplification procedures
Includes Stand
Quantity 1 Stand