missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega pGL4 Minimal Promoter Driven Luciferase Vectors
Firefly luciferase reporter vectors with response elements.
$831.40 - $832.72
Specifications
| Marker Gene | Hygro |
|---|---|
| Promoter | SV40 |
| Protein Degradation Sequence | hPEST |
| Reporter Gene | luc2P |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 1mM EDTA |
| Catalog Number | Mfr. No. | Vector | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Vector | Price | Quantity | |||||
PRE3651
|
Promega
E3651 |
pGL4.38[luc2P/p53 RE/Hygro] |
Each for $831.40
|
|
|||||
PRE3751
|
Promega
E3751 |
pGL4.41[luc2P/HSE/Hygro] |
Each for $832.72
|
|
|||||
PRE3671
|
Promega
E3671 |
pGL4.48[luc2P/SBE/Hygro] |
Each for $831.40
|
|
|||||
PRE4041
|
Promega
E4041 |
pGL4.47[luc2P/SIE/Hygro] |
Each for $831.40
|
|
|||||
PRE4131
|
Promega
E4131 |
pGL4.40[luc2P/MRE/Hygro] |
Each for $831.40
|
|
|||||
PRE4141
|
Promega
E4141 |
pGL4.45[luc2P/ISRE/Hygro] |
Each for $831.40
|
|
|||||
PRE4611
|
Promega
E4611 |
pGL4.49[luc2P/TCF-LEFRE/Hygro] |
Each for $832.72
|
|
|||||
PRE3661
|
Promega
E3661 |
pGL4.39[luc2P/ATF6 RE/Hygro] |
Each for $831.40
|
|
|||||
Description
- Offer a range of response elements that are involved in stress, cytokine, nuclear response, kinase and other signaling pathways
- Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
| Hygro | |
| hPEST | |
| 10mM Tris HCl (pH 7.4), 1mM EDTA | |
| 1 μg/μL | |
| 20 μg |
| SV40 | |
| luc2P | |
| -30°C to -10°C | |
| RV Primer 3 and RV Primer 4 | |
| Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis, transient transfection experiments |