Learn More
Promega pGL4 Luciferase Reporter Vectors
Available in numerous configurations for enhanced usability and convenience
Supplier:  Promega E8481
Encompass
 View more versions of this product
Catalog No. PRE8481
Only null left
Description
Description
  • Reporter Vectors Ready to Use in Your Signaling Pathway Without Cloning
  • Offer a number of response elements that are involved in nuclear response, kinase and other signaling pathways
  • Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
Chemical Identifiers
-30°C to -10°C
1 μg/μL
20 μg
Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Specifications
Hygro
hPEST
10mM Tris HCl pH 7.4, 1mM EDTA
1 μg/μL
20 μg
Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
NFAT-RE
luc2A
-30°C to -10°C
RV Primer 3 and RV Primer 4
pGL4.30[luc2P/NFAT-RE/Hygro]
Product Suggestions
Product Suggestions