missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega pGL4 Luciferase Reporter Vectors
Available in numerous configurations for enhanced usability and convenience
Supplier: Promega E8421
Description
- Vectors Used for Cloning a Response Element Upstream of Firefly Luciferase Gene
- Minimal promoter (minP) helps drive expression of luciferase when a response element is cloned upstream
- Available with three varieties of engineered firefly luciferase genes: luc2, luc2P or luc2CP
Chemical Identifiers
| -30°C to -10°C | |
| 1 μg/μL | |
| 20 μg | |
| Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis |
Specifications
| minP | |
| luc2A | |
| -30°C to -10°C | |
| RV Primer 3 and RV Primer 4 | |
| pGL4.24[luc2P/minP] |
| hPEST | |
| 10mM Tris HCl pH 7.4, 1mM EDTA | |
| 1 μg/μL | |
| 20 μg | |
| Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis |