missing translation for 'onlineSavingsMsg'
Learn More

Promega pGL4 Luciferase Reporter Vectors

Catalog No. PRE1360
Encompass
Change view
Click to view available options
Reporter Gene:
hRlucB
luc2A
Storage Buffer:
10mM Tris HCl pH 7.4, 1mM EDTA
10mM Tris HCl, 1mM EDTA (pH 8)
Vector:
pGL4.10[luc2]
pGL4.13[luc2/SV40]
pGL4.14[luc2/Hygro]
pGL4.15[luc2P/Hygro]
pGL4.17[luc2/Neo]
pGL4.18[luc2P/Neo]
pGL4.20[luc2/Puro]
pGL4.22[luc2CP/Puro]
pGL4.23[luc2/minP]
pGL4.24[luc2P/minP]
pGL4.25[luc2CP/minP]
pGL4.26[luc2/minP/Hygro]
pGL4.27[luc2P/minP/Hygro]
pGL4.28[luc2CP/minP/Hygro]
pGL4.29[luc2P/CRE/Hygro]
pGL4.30[luc2P/NFAT-RE/Hygro]
pGL4.31[luc2P/GAL4UAS/Hygro]
pGL4.32[luc2P/NF-kB-RE/Hygro]
pGL4.33[luc2P/SRE/Hygro]
pGL4.34[luc2P/SRF-RE/Hygro]
pGL4.35[luc2P/9XGAL4UAS/Hygro]
pGL4.36[luc2P/MMTV/Hygro]
pGL4.37[luc2P/ARE/Hygro]
pGL4.42[luc2P/HRE/Hygro]
pGL4.43[luc2P/XRE/Hygro]
pGL4.44[luc2P/AP1 RE/Hygro]
pGL4.50[ luc2 /CMV/Hygro]
pGL4.51[luc2/CMV/Neo]
pGL4.52[luc2P/STAT5RE/Hygro]
pGL4.70[hRluc]
pGL4.73[hRluc/SV40]
pGL4.74[hRluc/TK]
pGL4.75[hRluc/CMV]
34 product options available for selection
Product selection table with 34 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Reporter Gene Storage Buffer Vector
PRE1360 luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.36[luc2P/MMTV/Hygro]
PRE6651 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.10[luc2]
PRE6661 - - -
PRE6681 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.13[luc2/SV40]
PRE6721 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.17[luc2/Neo]
PRE6731 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.18[luc2P/Neo]
PRE6911 hRlucB 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.73[hRluc/SV40]
PRE6771 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.22[luc2CP/Puro]
PRE6881 hRlucB 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.70[hRluc]
PRE6921 hRlucB 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.74[hRluc/TK]
PRE6931 hRlucB 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.75[hRluc/CMV]
PRE6751 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.20[luc2/Puro]
PRE6701 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.15[luc2P/Hygro]
PRE6691 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.14[luc2/Hygro]
PRE8491 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.32[luc2P/NF-kB-RE/Hygro]
PRE1340 luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.33[luc2P/SRE/Hygro]
PRE1350 luc2A 10mM Tris HCl, 1mM EDTA (pH 8) pGL4.34[luc2P/SRF-RE/Hygro]
PRE1370 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.35[luc2P/9XGAL4UAS/Hygro]
P3935A luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.31[luc2P/GAL4UAS/Hygro]
PRE1310 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.50[ luc2 /CMV/Hygro]
PRE1320 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.51[luc2/CMV/Neo]
PRE8411 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.23[luc2/minP]
PRE8431 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.25[luc2CP/minP]
PRE8441 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.26[luc2/minP/Hygro]
PRE8421 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.24[luc2P/minP]
PRE8451 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.27[luc2P/minP/Hygro]
PRE8481 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.30[luc2P/NFAT-RE/Hygro]
PRE8461 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.28[luc2CP/minP/Hygro]
PRE8471 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.29[luc2P/CRE/Hygro]
PRE3641 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.37[luc2P/ARE/Hygro]
PRE4001 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.42[luc2P/HRE/Hygro]
PRE4121 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.43[luc2P/XRE/Hygro]
PRE4111 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.44[luc2P/AP1 RE/Hygro]
PRE4651 luc2A 10mM Tris HCl pH 7.4, 1mM EDTA pGL4.52[luc2P/STAT5RE/Hygro]
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 34 options available.
34 options
Catalog No. PRE1360 Supplier Promega Supplier No. E1360
Only null left

Available in numerous configurations for enhanced usability and convenience

  • Vectors To Bind Nuclear Receptors Using Recognition Sequences or One-Hybrid Assay
  • Create a binding domain fusion with the pFN26 (BIND) hRluc-neo Flexi(R) Vector and use with the pGL4.35[luc2P/9XGAL4UAS/Hygro] Vector
  • Mouse mammary tumor virus long terminal repeat in pGL4.36[luc2P/MMTV/Hygro] Vector assesses androgen or glucocorticoid responses
  • The estrogen receptor ligand binding domain (pBIND-Er∝) and the glucocorticoid receptor ligand binding domain (pBIND-GR) are already cloned

  • For research use only. This product and/or its use is subject to one or more of the following Promega patents: U.S. Pat. Appln. Ser. Nos. 09/645,706; 10/664,341; 10/943,508; U.S. Patents 5,670,356; 6,387,675 and 6,552,179, Australian Patent 698424 and various corresponding patent applications and issued patents. The terms of the limited license conveyed with the purchase of this product are as follows: Researchers may use this product in their own research and they may transfer derivatives to others for such research use provided that at the time of transfer a copy of this label license is given to the recipients and the recipients agree to be bound by the conditions of this label license. Researchers shall have no right to modify or otherwise create variations of the nucleotide sequence of the luciferase gene except that Researchers may: (1) clone heterologous DNA sequences at either or both ends of said luciferase gene so as to create fused gene sequences provided that the coding sequence of the resulting luciferase gene has no more than four deoxynucleotides missing at the affected terminus when compared to the intact luciferase gene sequence, and (2) insert and remove nucleic acid sequences in furtherance of splicing research predicated on the inactivation or reconstitution of the luminescent activity of the encoded luciferase. In addition, Researchers must do one of the following: (1) use luminescent assay reagents purchased from Promega Corporation for all determinations of luminescence activity resulting from the research use of this product and its derivatives; or (2) contact Promega to obtain a license for the use of the product and its derivatives. No other use or transfer of this product or its derivatives is authorized without the express written consent of Promega including, without limitation, Commercial Use. Commercial Use means any and all uses of this product and derivatives by a party for monetary or other consideration and may include but is not limited to use in: (1) product manufacture; and (2) to provide a service, information or data; and/or resale of the product or its derivatives, whether or not such product or derivatives are resold for use in research. With respect to such Commercial Use, or any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. If the purchaser is not willing to accept the conditions of this limited use statement, Promega is willing to accept the return of the unopened product and provide the purchaser with a full refund. However, in the event the product is opened, then the purchaser agrees to be bound by the conditions of this limited use statement.
  • Mfr. No. C9351, E1310, E1320, E1370, E6651, E6661, E6671, E6681, E6691, E6701, E6711, E6721, E6731, E6741, E6751, E6761, E6771, E8411, E8421, E8431, E8441, E8451, E8461, E8471, E8481, E8491: The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Mfr. No. E6881, E6891, E6901, E6911, E6921, E6931, E6941, E6951, E6961, E6971, E6981, E6991, E7501, E7511, E7521: Licensed from University of Georgia Research Foundation, Inc., under U.S. Patent 5,418,155; Canadian Patents 2,105,984 and related patents.
  • Mfr. No. E8481: The NFAT response element and its use are licensed under one or more of the following patents: U.S. Patents 5,837,840; 6,197,925; 5,989,810; 6,096,515; 6,388,052; 6,150,099; 6,171,781; 6,352,830; 6,312,899 and 6,875,571.
  • Mfr. No. E1310, E1320: The use of this product and derivatives thereof is strictly limited to that of a life sciences research reagent. All other use is strictly prohibited, including but not limited to any diagnostic, th

Specifications

Marker Gene Hygro
Promoter MMTV
Protein Degradation Sequence hPEST
Reporter Gene luc2A
Storage Buffer 10mM Tris HCl, 1mM EDTA (pH 8)
Content And Storage -30°C to -10°C
Concentration 1 μg/μL
Primer RV Primer 3 and RV Primer 4
Quantity 20 μg
Vector pGL4.36[luc2P/MMTV/Hygro]
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis
Show More Show Less